ω-Bromo-3-methoxyacetophenone ≥98%

Synonyms: 2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone

115673-10G 115673-25G 115673-1G
220034-537EA 107.48 USD
220034-537 220034-539 220034-538
ω-Bromo-3-methoxyacetophenone ≥98%
ω-Bromo-3-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 63…64.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000199
CAS Number: 5000-65-7
EINECS: 225-666-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR