ω-Bromo-4-methoxyacetophenone ≥97%

Warning

Synonyms: 4-Methoxyphenacyl bromide, Bromomethyl p-methoxyphenyl ketone, α-Bromo-4-methoxyacetophenone, 2-Bromo-1-(4-methoxyphenyl)ethan-1-one, 2-Bromo-1-(4-methoxyphenyl)ethanone, 2-Bromo-4'-methoxyacetophenone

115665-5G 115665-25G 115665-100G
220034-536EA 86.69 USD
220034-536 220034-535 220034-534
ω-Bromo-4-methoxyacetophenone ≥97%
ω-Bromo-4-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000201
CAS Number: 2632-13-5
EINECS: 220-118-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR