5-Chloro-2-nitrobenzonitrile ≥97%

115614-10G 115614-5G
220034-527EA 62.77 USD
220034-527 220034-528
5-Chloro-2-nitrobenzonitrile ≥97%
5-Chloro-2-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Boiling Pt: 120 °C (20 mmHg)
Melting Pt: 97 °C
Density: 1.584 g/cm³ (20 °C)
MDL Number: MFCD00007286
CAS Number: 34662-31-2
EINECS: 252-132-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR