(R)-(+)-α-Methylbenzylamine ≥98%

Danger

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

115541-10ML 115541-100ML 115541-25ML
220034-516EA 75.71 USD
220034-516 220034-515 220034-517
(R)-(+)-α-Methylbenzylamine ≥98%
(R)-(+)-α-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 32.5 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
Merck Index: 12,06106

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR