2'-Hydroxy-5'-methoxyacetophenone ≥99%

Synonyms: 2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone

114995-25G 114995-5G
220033-997EA 162.62 USD
220033-997 220033-998
2'-Hydroxy-5'-methoxyacetophenone ≥99%
2'-Hydroxy-5'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 113 °C
MDL Number: MFCD00008731
CAS Number: 705-15-7
EINECS: 211-882-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR