Phenylhydrazinium chloride ≥99%

Danger

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

114715-100G 114715-5G 114715-500G
220033-971EA 133.54 USD
220033-971 220033-973 220033-972
Phenylhydrazinium chloride ≥99%
Phenylhydrazinium chloride
Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: 243…246 °C
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1,III
Merck Index: 12,07447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR