Thymolphthalein ACS

Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

114553-10G 114553-50G
220033-956EA 51.66 USD
220033-956 220033-957
Thymolphthalein ACS
Thymolphthalein
Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
EINECS: 204-729-7
Merck Index: 12,09542

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR