Thymol blue ACS

Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

114545-25G 114545-5G
220033-954EA 200.2 USD
220033-954 220033-955
Thymol blue ACS
Thymol blue
Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR