Phenol red sodium salt ACS

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

114537-25G 114537-5G
220033-952EA 154 USD
220033-952 220033-953
Phenol red sodium salt ACS
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR