Methyl orange ACS

Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

114510-100G 114510-500G 114510-25G
220033-945EA 101.35 USD
220033-945 220033-947 220033-946
Methyl orange ACS
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: > 300 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 13,06127
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=915f043445c2be992ab36bc290d60762365a46ad&vid=bd67cfd7651199e11bfc8312958675f5db23665e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR