Methyl red sodium salt ACS

Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate

114502-100G 114502-25G
220034-468EA 200.2 USD
220034-468 220034-469
Methyl red sodium salt ACS
Methyl red sodium salt
Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Melting Pt: ? 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR