Bromothymol blue sodium salt ACS

Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

114421-5G 114421-25G
220034-462EA 54.05 USD
220034-462 220034-461
Bromothymol blue sodium salt ACS
Bromothymol blue sodium salt
Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
Melting Pt: ? 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f354f58906e22f2bb240fbb7582547d19079330&vid=5b5d658a2b6164adbb36ca73400e0f81bd70054a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR