Bromothymol blue ACS

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

114413-25G 114413-5G
220034-459EA 169.4 USD
220034-459 220034-460
Bromothymol blue ACS
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Melting Pt: 204…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR