Bromophenol blue sodium salt

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

114405-50G 114405-5G 114405-25G
220034-457EA 263.2 USD
220034-457 220034-458 220034-456
Bromophenol blue sodium salt
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba72d9d24b4b3189026592506f4645642ce22624&vid=dc56d7237e0f5fe0ec6698490819686f756bd39b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR