Bromocresol green sodium salt ACS

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

114367-5G 114367-25G
220034-193EA 91 USD
220034-193 220034-192
Bromocresol green sodium salt ACS
Bromocresol green sodium salt
Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6576f0c57895b913ac2b0cdaad5ae705df1a91ad&vid=e5e334727f1a16bf4bbcf034a9586a032e897bd9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR