Bromocresol green ACS

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

114359-25G 114359-5G
220034-190EA 281.4 USD
220034-190 220034-191
Bromocresol green ACS
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR