3,5-Diaminobenzoic acid dihydrochloride ≥99%

113832-25G 113832-100G
220034-444EA 523.94 USD
220034-444 220034-442
3,5-Diaminobenzoic acid dihydrochloride ≥99%
3,5-Diaminobenzoic acid dihydrochloride
Formula: C₇H₁₀ClN₂O₂
MW: 189.62 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00012983
CAS Number: 618-56-4
EINECS: 210-556-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR