2-Amino-3-nitropyridine

Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

113514-25G 113514-5G
220033-866EA 216.46 USD
220033-866 220033-867
2-Amino-3-nitropyridine
2-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 166…168 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR