6-Nitro-m-toluic acid ≥95%

Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

113409-25G 113409-5G
220033-862EA 174.46 USD
220033-862 220033-863
6-Nitro-m-toluic acid ≥95%
6-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 137 - 138 °C
MDL Number: MFCD00007368
CAS Number: 3113-72-2
EINECS: 221-481-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR