1,3-Diisopropylbenzene ≥96%

113263-500G 113263-100G
220033-282EA 132.46 USD
220033-282 220033-281
1,3-Diisopropylbenzene ≥96%
1,3-Diisopropylbenzene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: -40 °C
Density: 0.85593 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008889
CAS Number: 99-62-7
EINECS: 202-773-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR