Octylbenzene ≥98%

Synonyms: 1-Phenyloctane

113190-25G 113190-100G 113190-5G
220033-203EA 119.54 USD
220033-203 220033-202 220033-204
Octylbenzene ≥98%
Octylbenzene
Formula: C₁₄H₂₂
MW: 190.33 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.8562 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009564
CAS Number: 2189-60-8
EINECS: 218-582-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR