Pentylbenzene ≥99%

Synonyms: 1-Phenylpentane, Amylbenzene

113174-25G 113174-100G
220033-517EA 97.89 USD
220033-517 220033-516
Pentylbenzene ≥99%
Pentylbenzene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: -1 °C
Density: 0.8594 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009502
CAS Number: 538-68-1
EINECS: 208-701-5
UN: 3082
ADR: 9,III
Merck Index: 13,00604

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR