4-Methoxybenzyl mercaptan ≥90%, Technical Grade

Synonyms: 4-Methoxy-α-toluenethiol, p-Methoxytoluene-α-thiol

113158-25G 113158-5G
220034-435EA 393.08 USD
220034-435 220033-513
4-Methoxybenzyl mercaptan ≥90%, Technical Grade
4-Methoxybenzyl mercaptan
Formula: C₈H₁₀OS
MW: 154.23 g/mol
Boiling Pt: 90…95 °C (0.67 hPa)
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004871
CAS Number: 6258-60-2
EINECS: 228-393-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR