Oxalyl dibromide ≥97%

113034-25G 113034-100G
220034-427EA 130.85 USD
220034-427 220034-426
Oxalyl dibromide ≥97%
Oxalyl dibromide
Formula: C₂Br₂O₂
MW: 215.83 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 19.5 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000113
CAS Number: 15219-34-8
EINECS: 239-271-7
UN: 2922
ADR: 8,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR