2,5-Di-tert-butylhydroquinone ≥99%

Synonyms: BHQ

112976-25G 112976-1KG
220034-417EA 34.89 USD
220034-417 220034-416
2,5-Di-tert-butylhydroquinone ≥99%
2,5-Di-tert-butylhydroquinone
Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 212 °C
Density: 1.078 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008825
CAS Number: 88-58-4
EINECS: 201-841-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR