2-tert-Butylhydroquinone ≥97%

Synonyms: t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ

112941-100G 112941-500G 112941-5G
220034-411EA 53.62 USD
220034-411 220034-412 220034-413
2-tert-Butylhydroquinone ≥97%
2-tert-Butylhydroquinone
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 128 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002344
CAS Number: 1948-33-0
EINECS: 217-752-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR