Diethyl succinate ≥99%

Synonyms: Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester

112402-1KG 112402-250G
220034-305EA 141.23 USD
220034-305 220034-306
Diethyl succinate ≥99%
Diethyl succinate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR