Ethyl octanoate ≥99%

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

112321-5G 112321-100G
220034-294EA 17.88 USD
220034-294 220034-293
Ethyl octanoate ≥99%
Ethyl octanoate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR