4-Nitrobenzoyl chloride ≥98%

Danger

Synonyms: p-Nitrobenzoyl chloride

112208-250G 112208-50G 112208-500G
220034-274EA 97.03 USD
220034-274 220034-276 220034-275
4-Nitrobenzoyl chloride ≥98%
4-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 97 - 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8,II
Merck Index: 12,06687

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR