α,4-Dibromotoluene ≥98%

Synonyms: 4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene

112186-100G 112186-25G
220033-260EA 75.38 USD
220033-260 220033-261
α,4-Dibromotoluene ≥98%
α,4-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 129…130 °C (18 mmHg)
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,01393

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR