2-Methylbenzothiazole

112143-25G 112143-100G
220033-254EA 61.06 USD
220033-254 220033-253
2-Methylbenzothiazole
2-Methylbenzothiazole
Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.173 g/cm³ (20 °C)
MDL Number: MFCD00005794
CAS Number: 120-75-2
EINECS: 204-423-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR