(±)-2-Methyl-2,4-pentanediol ≥99%

Warning

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol

112100-18KG 112100-3KG 112100-500G
220033-312EA 364 USD
220033-312 220033-313 220033-314
(±)-2-Methyl-2,4-pentanediol ≥99%
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.923 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR