3,4-Dichlorobenzoyl chloride ≥97%

Warning

111945-100G 111945-25G
220034-263EA 88.85 USD
220034-263 220034-264
3,4-Dichlorobenzoyl chloride ≥97%
3,4-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00000672
CAS Number: 3024-72-4
EINECS: 221-177-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR