2,4-Dichlorobenzoyl chloride ≥98%

Danger

Synonyms: 2,4-Dichlorobenzene-1-carbonyl chloride

111937-100G 111937-5G
220034-261EA 105.97 USD
220034-261 220034-262
2,4-Dichlorobenzoyl chloride ≥98%
2,4-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00000661
CAS Number: 89-75-8
EINECS: 201-936-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR