Propylcyclohexane ≥99%

Synonyms: 1-Cyclohexylpropane

111856-25G 111856-100G
220034-244EA 430.77 USD
220034-244 220034-243
Propylcyclohexane ≥99%
Propylcyclohexane
Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: -94.9 °C
Density: 0.7936 g/cm³ (20 °C)
MDL Number: MFCD00013770
CAS Number: 1678-92-8
EINECS: 216-836-6
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR