1-Bromo-4-ethylbenzene ≥97%

Synonyms: 4-(Bromophenyl)ethane

111414-10G 111414-50G
220034-162EA 66.69 USD
220034-162 220034-163
1-Bromo-4-ethylbenzene ≥97%
1-Bromo-4-ethylbenzene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: -43.5 °C
Density: 1.34226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR