Butyl vinyl ether ≥98%

Synonyms: n-Butyl vinyl ether

110299-1L 110299-500ML 110299-2L 110299-100ML
220034-107EA 123.45 USD
220034-107 220034-109 220034-108 220034-106
Butyl vinyl ether ≥98%
Butyl vinyl ether
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –91.9 °C
Density: 0.7804 g/cm³ (20 °C)
MDL Number: MFCD00009454
CAS Number: 111-34-2
EINECS: 203-860-7
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR