Acrylonitrile ≥99% stabilized

Danger

110213-18L 110213-4L 110213-1L 110213-2L 110213-5ML 110213-100ML 110213-500ML
220034-081EA 542.69 USD
220034-081 220034-084 220034-082 220034-083 220034-086 220034-080 220034-085
Acrylonitrile ≥99% stabilized
Acrylonitrile
Formula: C₃H₃N
MW: 53.06 g/mol
Boiling Pt: 77.3 °C (1013 hPa)
Melting Pt: -83.55 °C
Density: 0.7951 g/cm³ (20 °C)
Flash Pt: -5 °C
MDL Number: MFCD00001927
CAS Number: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: 3,I
Merck Index: 13,00133

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR