DL-Arginine ≥97.0%

Synonyms: Arginine, (±)-Arginine, H-DL-Arg-OH, H-Arg-OH

11020-25G 11020-5G
220034-073EA 161.54 USD
220034-073 220034-074
DL-Arginine ≥97.0%
DL-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174.20 g/mol
MDL Number: MFCD00063117
CAS Number: 7200-25-1
EINECS: 230-571-3

Order Now

Specification Test Results

Assay 97.0% (NT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR