1,2-Epoxybutane ≥99%

Synonyms: 1,2-Butylene oxide, 1,2-Butylene Oxide, 1,2-Epoxybutane, Ethylethylene Oxide, Ethyloxirane

109975-250ML 109975-1L
220034-047EA 84.48 USD
220034-047 220034-046
1,2-Epoxybutane ≥99%
1,2-Epoxybutane
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: -130 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: -26 °C
Storage Temperature: Ambient
MDL Number: MFCD00005153
CAS Number: 106-88-7
EINECS: 203-438-2
UN: 3022
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR