2-Ethoxyethyl acetate ≥98%

Danger

Synonyms: Ethylglycol acetate

109967-25ML 109967-4L 109967-1L
220034-044EA 28.22 USD
220034-044 220034-045 220034-043
2-Ethoxyethyl acetate ≥98%
2-Ethoxyethyl acetate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.976 g/cm³ (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00009251
CAS Number: 111-15-9
EINECS: 203-839-2
UN: 1172
ADR: 3,III
Merck Index: 13,03787

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR