(±)-4-Methyl-2-pentanol ≥98%

Danger

Synonyms: (±)-Methyl isobutyl carbinol

109916-1L 109916-4L 109916-100ML
220033-246EA 46.85 USD
220033-246 220033-247 220033-245
(±)-4-Methyl-2-pentanol ≥98%
(±)-4-Methyl-2-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0.8093 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR