2-Methoxyethyl acetate ≥98%, Reagent Grade

Danger

Synonyms: Acetic acid 2-Methoxyethyl ester, Ethylene glycol monomethyl ether acetate, Methyl glycol acetate, Methyl Cellosolve acetate

109886-1L 109886-4L
220032-654EA 41.54 USD
220032-654 220033-244
2-Methoxyethyl acetate ≥98%, Reagent Grade
2-Methoxyethyl acetate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00008719
CAS Number: 110-49-6
EINECS: 203-772-9
Merck Index: 12,06119

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR