Propylamine ≥98%

Danger

Synonyms: 1-Propylamine, 1-Aminopropane

109819-1L 109819-250ML
220033-933EA 79.05 USD
220033-933 220033-934
Propylamine ≥98%
Propylamine
Formula: H₃CCH₂CH₂NH₂
MW: 59.11 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: -83 °C
Density: 0.7175 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008205
CAS Number: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3,II
Merck Index: 12,08028

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR