2-Amino-6-chloropurine

Synonyms: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

109789-5G 109789-1G
220033-930EA 21.54 USD
220033-930 220033-929
2-Amino-6-chloropurine
2-Amino-6-chloropurine
Formula: C₅H₄ClN₅
MW: 169.57 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR