4-(Dimethylamino)benzaldehyde ≥98%

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

109762-100G 109762-500G
220033-927EA 60.85 USD
220033-927 220033-928
4-(Dimethylamino)benzaldehyde ≥98%
4-(Dimethylamino)benzaldehyde
Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176-177 °C (1013 hPa)
Melting Pt: 72 °C
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR