N-Chlorosuccinimide

Danger

Synonyms: 1-Chloropyrrolidine-2,5-dione

109681-100G 109681-500G
220033-921EA 34.25 USD
220033-921 220033-922
N-Chlorosuccinimide
N-Chlorosuccinimide
Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Boiling Pt: 216.5 °C (1013 hPa)
Melting Pt: 148 - 151 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 13,02183

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR