1-Hydroxy-2-naphthoic acid

Warning

Synonyms: 1-Hydroxy-naphthalene-2-carboxylic acid

109630-100G 109630-5G
220033-918EA 61.6 USD
220033-918 220033-919
1-Hydroxy-2-naphthoic acid
1-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 205 °C
MDL Number: MFCD00003960
CAS Number: 86-48-6
EINECS: 201-674-0
UN: 2811
ADR: 6.1,III
Merck Index: 13,04859

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR