o-Anisaldehyde ≥98%

Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde

109622-100G 109622-500G 109622-2G
220033-915EA 64.62 USD
220033-915 220033-917 220033-916
o-Anisaldehyde ≥98%
o-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 34…37 °C
Density: 1.127 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4
EINECS: 205-171-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR