Dimethyl itaconate

Synonyms: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

109533-100G 109533-500G
220033-905EA 71.94 USD
220033-905 220033-906
Dimethyl itaconate
Dimethyl itaconate
Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1.1399 g/cm³ (14.7 °C)
MDL Number: MFCD00008446
CAS Number: 617-52-7
EINECS: 210-519-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR